missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA polymerase sigma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | DNA polymerase sigma |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18420282
|
Novus Biologicals
NBP2-13728-25ul |
25ul |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18096138
|
Novus Biologicals
NBP2-13728 |
0.1 mL |
£488.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA polymerase sigma Polyclonal antibody specifically detects DNA polymerase sigma in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
| DNA polymerase sigma | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| DNA polymerase kappa, DNA polymerase sigma, EC 2.7.7.7, LAK-1TUTase 5, PAP associated domain containing 7, POLKTRF41, POLSTUTASE5, polymerase (DNA directed) sigma, polymerase (DNA-directed) sigma, Terminal uridylyltransferase 5, Topoisomerase-related function protein 4-1, TRF4-1LAK1, TRF4PAP-associated domain-containing protein 7 | |
| TENT4A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 11044 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: PNPLSSPHLYHKQHNGMKLSMKGSHGHTQGGGYSSVGSGGVRPPVGNRGHHQYNRTGWRRKKHTHTRDSLPVSLSR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title