missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79290
This item is not returnable.
View return policy
Description
DNA Polymerase gamma Polyclonal specifically detects DNA Polymerase gamma in Rat samples. It is validated for Western Blot.
Specifications
| DNA Polymerase gamma | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5428 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_445980 | |
| POLG | |
| The specific Immunogen is proprietary information. Peptide sequence QDWQEQLVVGHNVSFDRAHIREQYLIQGSRMHFLDTMSMHMAISGLSSFQ. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Canine: 92%; Guinea pig: 92%; Equine: 92%; Zebrafish: 92%; Pig: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Yeast, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction