missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase epsilon p59 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£294.00 - £419.00
Specifications
| Antigen | DNA Polymerase epsilon p59 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18470882
|
Novus Biologicals
NBP1-87970-25ul |
25 μL |
£294.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18058003
|
Novus Biologicals
NBP1-87970 |
0.1 mL |
£419.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA Polymerase epsilon p59 Polyclonal specifically detects DNA Polymerase epsilon p59 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DNA Polymerase epsilon p59 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| DNA polymerase epsilon subunit 2, DNA polymerase II subunit 2, DPE2DNA polymerase epsilon subunit B, EC 2.7.7.7, polymerase (DNA directed), epsilon 2 (p59 subunit) | |
| POLE2 | |
| IgG | |
| Affinity Purified | |
| Specificity of human DNA Polymerase epsilon p59 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5427 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NAVEKQPLSSNMIERSVVEAAVQECSQSVDETIEHVFNIIGAFDIPRFVYNSERKKFLPLLMTNHPAPNLFGTPRDKAEMFRERYTILHQRTHRHELFTPPVIGSHPDESGSKFQLKTIETLLGSTTKIGDAIVLG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title