missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase epsilon p59 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-87970
This item is not returnable.
View return policy
Description
DNA Polymerase epsilon p59 Polyclonal specifically detects DNA Polymerase epsilon p59 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DNA Polymerase epsilon p59 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DNA polymerase epsilon subunit 2, DNA polymerase II subunit 2, DPE2DNA polymerase epsilon subunit B, EC 2.7.7.7, polymerase (DNA directed), epsilon 2 (p59 subunit) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human DNA Polymerase epsilon p59 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| POLE2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NAVEKQPLSSNMIERSVVEAAVQECSQSVDETIEHVFNIIGAFDIPRFVYNSERKKFLPLLMTNHPAPNLFGTPRDKAEMFRERYTILHQRTHRHELFTPPVIGSHPDESGSKFQLKTIETLLGSTTKIGDAIVLG | |
| 0.1 mL | |
| Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
| 5427 | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction