missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase delta, catalytic subunit Rabbit anti-Human, Mouse, Rat, Clone: 4B6I3, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | DNA Polymerase delta, catalytic subunit |
|---|---|
| Clone | 4B6I3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18386554
|
Novus Biologicals
NBP3-16358-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18314723
|
Novus Biologicals
NBP3-16358-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA Polymerase delta, catalytic subunit Monoclonal antibody specifically detects DNA Polymerase delta, catalytic subunit in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DNA Polymerase delta, catalytic subunit | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5424 | |
| IgG | |
| Affinity purified |
| 4B6I3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CDC2, CDC2 homolog, DNA polymerase delta catalytic subunit, DNA polymerase subunit delta p125, DNA-directed polymerase delta 1, EC 2.7.7.7, POLD, polymerase (DNA directed), delta 1, catalytic subunit (125kD), polymerase (DNA directed), delta 1, catalytic subunit 125kDa | |
| A synthetic peptide corresponding to a sequence within amino acids 1000-1107 of human DNA Polymerase delta, catalytic subunit (P28340). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title