missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase delta, catalytic subunit Rabbit anti-Human, Mouse, Rat, Clone: 4B6I3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16358-100UL
This item is not returnable.
View return policy
Description
DNA Polymerase delta, catalytic subunit Monoclonal antibody specifically detects DNA Polymerase delta, catalytic subunit in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DNA Polymerase delta, catalytic subunit | |
| Monoclonal | |
| Unconjugated | |
| CDC2, CDC2 homolog, DNA polymerase delta catalytic subunit, DNA polymerase subunit delta p125, DNA-directed polymerase delta 1, EC 2.7.7.7, POLD, polymerase (DNA directed), delta 1, catalytic subunit (125kD), polymerase (DNA directed), delta 1, catalytic subunit 125kDa | |
| A synthetic peptide corresponding to a sequence within amino acids 1000-1107 of human DNA Polymerase delta, catalytic subunit (P28340). | |
| 100 μg | |
| Breast Cancer, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
| 5424 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 4B6I3 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction