missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase beta Rabbit anti-Human, Mouse, Rat, Clone: 6M8I5, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | DNA Polymerase beta |
|---|---|
| Clone | 6M8I5 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18316465
|
Novus Biologicals
NBP3-16127-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18306702
|
Novus Biologicals
NBP3-16127-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA Polymerase beta Monoclonal antibody specifically detects DNA Polymerase beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| DNA Polymerase beta | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DNA pol beta, DNA polymerase beta, DNA polymerase beta subunit, EC 2.7.7.7, EC 4.2.99.-, MGC125976, polymerase (DNA directed), beta | |
| A synthetic peptide corresponding to a sequence within amino acids 236-335 of human DNA Polymerase beta (P06746). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 6M8I5 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA Polymerases, DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5423 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title