missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase beta Rabbit anti-Human, Mouse, Rat, Clone: 6M8I5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16127-100UL
This item is not returnable.
View return policy
Description
DNA Polymerase beta Monoclonal antibody specifically detects DNA Polymerase beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DNA Polymerase beta | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6M8I5 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| DNA pol beta, DNA polymerase beta, DNA polymerase beta subunit, EC 2.7.7.7, EC 4.2.99.-, MGC125976, polymerase (DNA directed), beta | |
| A synthetic peptide corresponding to a sequence within amino acids 236-335 of human DNA Polymerase beta (P06746). | |
| 100 μg | |
| DNA Polymerases, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 5423 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction