missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA-PKcs Antibody (3H6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£187.00 - £400.00
Specifications
| Antigen | DNA-PKcs |
|---|---|
| Clone | 3H6 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18176661
|
Novus Biologicals
NBP2-22128SS |
0.025 mL |
£187.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18750214
|
Novus Biologicals
NBP2-22128 |
£400.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
DNA-PKcs Monoclonal antibody specifically detects DNA-PKcs in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
| DNA-PKcs | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, DNA replication Transcription Translation and Splicing, Non homologous end joining, Phospho Specific, Protein Kinase | |
| PBS with 0.02% Sodium Azide | |
| DNA-dependent protein kinase, DNA-dependent protein kinase catalytic subunit-like, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, EC 2.7.11.1, HYRC, HYRC1, p460 | |
| PRKDC | |
| IgG1 | |
| Protein G purified | |
| 460 kDa |
| 3H6 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse | |
| P78527 | |
| 5591 | |
| Purified recombinant fragment of human DNA PKcs expressed in E. coli. [Uniprot: P78527] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title