missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA-PKcs Antibody (3H6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22128SS
This item is not returnable.
View return policy
Description
DNA-PKcs Monoclonal antibody specifically detects DNA-PKcs in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| DNA-PKcs | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| P78527 | |
| PRKDC | |
| Purified recombinant fragment of human DNA PKcs expressed in E. coli. [Uniprot: P78527] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Ascites |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3H6 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DNA-dependent protein kinase, DNA-dependent protein kinase catalytic subunit-like, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, EC 2.7.11.1, HYRC, HYRC1, p460 | |
| Mouse | |
| 460 kDa | |
| 0.025 mL | |
| Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, DNA replication Transcription Translation and Splicing, Non homologous end joining, Phospho Specific, Protein Kinase | |
| 5591 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction