missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ DNA LoBind™ Tubes

Product Code. 17453703 Shop All Eppendorf Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2.0 mL
5 mL
15 mL
25 mL
50 mL
Packaging:
1 Bag x 500 Tubes
200 Tubes (2 Bags x 100 Tubes)
200 Tubes (4 Bags x 50 Tubes)
200 Tubes (5 Bags x 40 Tubes)
200 Tubes (8 Bags x 25 Tubes)
4 Bags of 50
Closure Type:
Screw Cap
Snap Cap
SnapTec Cap
Max. RCF:
17,000 x g
19,500 x g
25,000 x g
30,000 x g
Unit Size:
Pack of 200
Pack of 500
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Packaging Closure Type Max. RCF unitSize
17453703 25 mL 200 Tubes (4 Bags x 50 Tubes) Screw Cap 17,000 x g Pack of 200
16618742 0.5 mL 1 Bag x 500 Tubes Snap Cap 30,000 x g Pack of 500
16628742 1.5 mL 1 Bag x 500 Tubes Snap Cap 30,000 x g Pack of 500
16638742 2.0 mL 1 Bag x 500 Tubes Snap Cap 25,000 x g Pack of 500
16170371 5 mL 200 Tubes (2 Bags x 100 Tubes) Screw Cap 25,000 x g Pack of 200
15571312 15 mL 200 Tubes (4 Bags x 50 Tubes) Screw Cap 19,500 x g Pack of 200
17423703 25 mL 200 Tubes (5 Bags x 40 Tubes) SnapTec Cap 17,000 x g Pack of 200
15581312 50 mL 200 Tubes (8 Bags x 25 Tubes) Screw Cap 19,500 x g Pack of 200
15168344 5 mL 4 Bags of 50 Snap Cap 25,000 x g Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 17453703 Supplier Eppendorf™ Supplier No. 0030122275
Only null left
Add to basket
Edge
Add to basket

Unleash the power of precision in your DNA research with Eppendorf DNA LoBind Tubes. Engineered with proprietary LoBind technology, these microcentrifuge tubes redefine sample handling, minimizing the risk of loss and maximizing DNA recovery. Ideal for low-concentration applications, including PCR, qPCR, and DNA sequencing, these tubes ensure accuracy, purity, and optimal yields.

Maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding with these tubes. Eppendorf™ Polypropylene DNA LoBind Tubes are ideal for sample preparation and long-term storage.

  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Precision lid sealing minimizes evaporation rates in the tube format.
  • Suitable for molecular biology applications with a centrifugation speed rating of up to 30,000 x g (25,000 x g for 2.0 mL tube).
  • Snap, screw, and SnapTec™ cap options available.
TRUSTED_SUSTAINABILITY

Specifications

Closure Material HDPE
Color Clear
Capacity (Metric) 25 mL
Material Polypropylene
Packaging 200 Tubes (4 Bags x 50 Tubes)
Quantity 200 Tubes (2 Bags of 100)
Sterility Non-sterile
Autoclavable Not Autoclavable
Shape Cylindrical
Clean Claims PCR Clean
Bottom Shape Conical
Closure Type Screw Cap
Product Type Tube
Max. RCF 17,000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.