missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMTF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | DMTF1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232656
|
Novus Biologicals
NBP3-35717-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228370
|
Novus Biologicals
NBP3-35717-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DMTF1 Polyclonal antibody specifically detects DMTF1 in Human samples. It is validated for ELISA,Western BlotSpecifications
| DMTF1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 9988 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cyclin D binding myb-like transcription factor 1, cyclin D-binding Myb-like protein 1, cyclin-D-binding Myb-like transcription factor 1, Cyclin-D-interacting Myb-like protein 1, DMP1FLJ76054, FLJ25188, FLJ41265, hDMP1DMTF, hDMTF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 561-760 of human DMTF1 (NP_066968.3).,, Sequence:, VTVQCHTPRVIIQTVATEDITSSISQAELTVDSDIQSSDFPEPPDALEADTFPDEIHHPKMTVEPSFNDAHVSKFSDQNSTELMNSVMVRTEEEISDTDLKQEESPSDLASAYVTEGLESPTIEEQVDQTIDDETILIVPSPHGFIQASDVIDTESVLPLTTLTDPILQHHQEESNIIGSSLGSPVSEDSKDVEDLVNCH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title