missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMTF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35717-20ul
This item is not returnable.
View return policy
Description
DMTF1 Polyclonal antibody specifically detects DMTF1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| DMTF1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| cyclin D binding myb-like transcription factor 1, cyclin D-binding Myb-like protein 1, cyclin-D-binding Myb-like transcription factor 1, Cyclin-D-interacting Myb-like protein 1, DMP1FLJ76054, FLJ25188, FLJ41265, hDMP1DMTF, hDMTF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 561-760 of human DMTF1 (NP_066968.3).,, Sequence:, VTVQCHTPRVIIQTVATEDITSSISQAELTVDSDIQSSDFPEPPDALEADTFPDEIHHPKMTVEPSFNDAHVSKFSDQNSTELMNSVMVRTEEEISDTDLKQEESPSDLASAYVTEGLESPTIEEQVDQTIDDETILIVPSPHGFIQASDVIDTESVLPLTTLTDPILQHHQEESNIIGSSLGSPVSEDSKDVEDLVNCH | |
| 20 μL | |
| Breast Cancer | |
| 9988 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction