missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMBT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DMBT1 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16825365
|
Proteintech
27069-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16815365
|
Proteintech
27069-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycoprotein-340 is member of the scavenger receptor superfamily and has been identified as a putative SP-D receptor. Gp-340 has been shown to bind SP-D in a calcium dependent manner through a protein-protein interaction. Like SP-D, gp-340 is mainly found in lung tissue and immunohistochemistry revealed high expression in alveolar macrophages. An identical protein is present in saliva (salivary agglutinin) and both forms are encoded by the gene DMBT1.Specifications
| DMBT1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UGM3 | |
| 1755 | |
| DMBT1 Fusion Protein Ag16229 | |
| Primary | |
| -20°C | |
| DMBT1 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DMBT1, Glycoprotein 340, Gp 340, GP340, Hensin, muclin, SAG, Salivary agglutinin, SALSA | |
| DMBT1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title