missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMAP1 Recombinant Rabbit Monoclonal Antibody (23GB5680), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | DMAP1 |
|---|---|
| Clone | 23GB5680 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088972
![]() |
Invitrogen™
MA553098 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088987
|
Invitrogen™
MA553099 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DMAP1 Recombinant Monoclonal antibody specifically detects DMAP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| DMAP1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9JI44, Q9NPF5 | |
| 298447, 55929, 66233 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5680 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1500016M21Rik; DKFZp686L09142; Dmap1; Dmtap1; DNA methyltransferase 1 associated protein 1; DNA methyltransferase 1-associated protein 1; DNMAP1; DNMT1 associated protein 1; DNMT1 associated protein-1; DNMT1-associated protein 1; DNMTAP1; EAF2; FLJ11543; KIAA1425; MAT1-mediated transcriptional repressor; MEAF2; mKIAA1425; Mmtr; RP5-891H21.2; SWC4 | |
| A synthesized peptide derived from human DMAP1 (411-467AA). | |
| Primary | |
| -20°C | |
| DMAP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title