missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLX3 Mouse anti-Human, Unconjugated, Clone: 1H8E1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DLX3 |
|---|---|
| Clone | 1H8E1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
Description
The DLX3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DLX3. This antibody recognizes human, mouse, rat, pig antigen. The DLX3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| DLX3 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 1747 | |
| Primary |
| 1H8E1 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Mouse, Rat, Pig | |
| IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title