Learn More
DLX3 Mouse anti-Human, Unconjugated, Clone: 1H8E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67282-1-IG-20UL
This item is not returnable.
View return policy
Description
The DLX3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DLX3. This antibody recognizes human, mouse, rat, pig antigen. The DLX3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| DLX3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, ELISA | |
| 1H8E1 | |
| Unconjugated | |
| 20 μL | |
| 1747 | |
| IgG2a |