missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLX3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33246-100ul
This item is not returnable.
View return policy
Description
DLX3 Monoclonal antibody specifically detects DLX3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| DLX3 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| AI4, distal-less homeo box 3, distal-less homeobox 3, homeobox protein DLX-3, TDO | |
| A synthetic peptide corresponding to a sequence within amino acids 188-287 of human DLX3 (O60479).,, Sequence:, YKNGEVPLEHSPNNSDSMACNSPPSPALWDTSSHSTPAPARSQLPPPLPYSASPSYLDDPTNSWYHAQNLSGPHLQQQPPQPATLHHASPGPPPNPGAVY | |
| 100 μL | |
| Growth and Development, Neuronal Cell Markers, Stem Cell Markers | |
| 1747 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction