missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL1 Rabbit anti-Human, Clone: 306, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | DLL1 |
|---|---|
| Clone | 306 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18260467
|
Novus Biologicals
NBP2-89936-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18227427
|
Novus Biologicals
NBP2-89936-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DLL1 Monoclonal specifically detects DLL1 in Human samples. It is validated for ELISA.Specifications
| DLL1 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
| Recombinant Human DLL1 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 306 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Stem Cell Signaling Pathway | |
| 0.2 um filtered solution in PBS | |
| 28514 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title