missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL1 Rabbit anti-Human, Clone: 306, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89936-100UL
This item is not returnable.
View return policy
Description
DLL1 Monoclonal specifically detects DLL1 in Human samples. It is validated for ELISA.
Specifications
| DLL1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 306 | |
| ELISA 1:5000-1:10000 | |
| Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
| Recombinant Human DLL1 protein | |
| 100 μg | |
| Neuroscience, Stem Cell Signaling Pathway | |
| 28514 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction