missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22474-0.025mg
This item is not returnable.
View return policy
Description
DLG1 Polyclonal specifically detects DLG1 in Rat samples. It is validated for Western Blot.
Specifications
| DLG1 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 1 mg/mL BSA with 0.05% Sodium Azide | |
| DLG1 | |
| Synthetic peptide corresponding to residues E(16) E Y R S K L S Q T E D R Q L R S S(33) of human SAP97. | |
| 0.025 mg | |
| Primary | |
| Rat | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 3 μg/mL | |
| discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1739 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction