missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00
Specifications
| Antigen | DLG1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 3 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18783194
|
Novus Biologicals
NBP2-22474-0.025mg |
0.025 mg | |||||||
Description
DLG1 Polyclonal specifically detects DLG1 in Rat samples. It is validated for Western Blot.Specifications
| DLG1 | |
| Western Blot 3 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Rat | |
| discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97 | |
| DLG1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS containing 1 mg/mL BSA with 0.05% Sodium Azide | |
| 1739 | |
| Synthetic peptide corresponding to residues E(16) E Y R S K L S Q T E D R Q L R S S(33) of human SAP97. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title