missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£174.00
Specifications
| Antigen | DLG1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DLG1 Polyclonal specifically detects DLG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| DLG1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Vision | |
| PBS with 0.05% Sodium Azide | |
| 1739 | |
| Synthetic peptide corresponding to residues V(115) L P S E R I S P Q V P N E V L G P E(133) of rat SAP97. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot 1:500 - 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97 | |
| DLG1 | |
| IgG | |
| Ammonium sulfate precipitation | |
| Detects SAP 97 from rat tissues. No cross-reactivity to other proteins, including synapse-associated protein family members, has been observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title