missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB600-1229-0.025ml
This item is not returnable.
View return policy
Description
DLG1 Polyclonal specifically detects DLG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| DLG1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:1000 | |
| discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Detects SAP 97 from rat tissues. No cross-reactivity to other proteins, including synapse-associated protein family members, has been observed. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| DLG1 | |
| Synthetic peptide corresponding to residues V(115) L P S E R I S P Q V P N E V L G P E(133) of rat SAP97. | |
| 0.025 mL | |
| Neuroscience, Vision | |
| 1739 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction