missing translation for 'onlineSavingsMsg'
Learn More

DL-Homocystine-3,3,3',3',4,4,4',4'-d8, TRC

Product Code. 30400401
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400401 25 mg
30399593 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400401 Supplier Toronto Research Chemicals Supplier No. TRCH59129225MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108641-82-3
Molecular Formula C8 D8 H8 N2 O4 S2
Molecular Weight (g/mol) 276.4029
Synonym (R*,R*)-(±)-4,4'-Dithiobis[2-amino-Butanoic Acid-d8, DL-4,4'-Dithiobis[2-amino-butyric Acid-d8, DL-4,4'-Dithiobis(2-aminobutyric acid)-d8, DL-Homocystine-d8, NSC 226570-d8
IUPAC Name rac-(2R)-2-amino-3,3,4,4-tetradeuterio-4-[[rac-(3R)-3-amino-3-carboxy-1,1,2,2-tetradeuteriopropyl]disulfanyl]butanoic acid
SMILES [2H]C([2H])(SSC([2H])([2H])C([2H])([2H])[C@H](N)C(=O)O)C([2H])([2H])[C@H](N)C(=O)O

Tekniske data

InChI Formula InChI=1S/C8H16N2O4S2/c9-5(7(11)12)1-3-15-16-4-2-6(10)8(13)14/h5-6H,1-4,9-10H2,(H,11,12)(H,13,14)/t5-,6-/m0/s1/i1D2,2D2,3D2,4D2
Quantity 25 mg
Formula Weight 276.1054
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material DL-Homocystine-3,3,3',3',4,4,4',4'-d8
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.