missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress DL-dithiothreitol

Product Code. 16409457
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16409457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16409457 Supplier MedChemExpress Supplier No. HY15917
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DL-dithiothreitol (DTT) is a reducing agent. DL-dithiothreitol forms a stable six-membered ring with an internal disulfide bond once oxidized.

Specifications

Molecular Weight (g/mol) 154.25 g/mol
CAS 12-3-3483
Content And Storage 4°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen)
Shipping Condition Room temperature
Quantity 5 g
Synonym DTT
Solubility H2O : ≥ 200 mg/mL (1296.60 mM) ∣ DMSO : 100 mg/mL (648.30 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.