missing translation for 'onlineSavingsMsg'
Learn More

DL-2,3-Diaminopropionic acid monohydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11357446
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357446 5 g 5g
11347446 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357446 Supplier Thermo Scientific Alfa Aesar Supplier No. L09485.06
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54897-59-5
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00012884
InChI Key SKWCZPYWFRTSDD-UHFFFAOYSA-N
Synonym 2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
PubChem CID 108638
IUPAC Name 2,3-diaminopropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)N.Cl

Specifications

Melting Point ∽232°C (decomposition)
Quantity 5 g
Beilstein 5293359
Merck Index 14,2984
Formula Weight 140.57
Percent Purity 99%
Chemical Name or Material DL-2,3-Diaminopropionic acid monohydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.