accessibility menu, dialog, popup

UserName

HClNH2NH2OHO

CAS RN 54897-59-5

IUPAC Name: hydrogen 2,3-diaminopropanoic acid chloride
Synonyms: hydrogen 2,3-diaminopropanoic acid chloride
Molecular Weight (g/mol): 140.57
Molecular Formula: C3H9ClN2O2
InChi Key: SKWCZPYWFRTSDD-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)