missing translation for 'onlineSavingsMsg'
Learn More

DL-2,3-Diaminopropionic acid monohydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number 10299403
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
54897-59-5
C3H8N2O2·HCl
140.57
MFCD00012884
SKWCZPYWFRTSDD-UHFFFAOYSA-N
2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
108638
2,3-diaminopropanoic acid;hydrochloride
C(C(C(=O)O)N)N.Cl
This item is not returnable. View return policy
54897-59-5
C3H8N2O2·HCl
140.57
MFCD00012884
SKWCZPYWFRTSDD-UHFFFAOYSA-N
2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
108638
2,3-diaminopropanoic acid;hydrochloride
C(C(C(=O)O)N)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 222°C to 226°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NCH2CH(NH2)CO2H·HCl
Quantity 25 g
Beilstein 04, 405
Merck Index 10, 2948
Solubility Information Solubility in water: freely soluble in water
Formula Weight 140.57
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-2, 3-Diaminopropionic acid monohydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • DL-2,3-Diaminopropionic acid monohydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'