missing translation for 'onlineSavingsMsg'
Learn More

DL-2,3-Diaminopropionic acid monohydrochloride, 99%

Product Code. 10062680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10062680 5 g Glass bottle 5g
10299403 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10062680 Supplier Thermo Scientific Acros Supplier No. 204670050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54897-59-5 | C3H8N2O2·HCl | 140.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54897-59-5
Molecular Formula C3H8N2O2·HCl
Molecular Weight (g/mol) 140.57
MDL Number MFCD00012884
InChI Key SKWCZPYWFRTSDD-UHFFFAOYSA-N
Synonym 2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
PubChem CID 108638
IUPAC Name 2,3-diaminopropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)N.Cl

Specifications

Melting Point 222°C to 226°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NCH2CH(NH2)CO2H·HCl
Quantity 5 g
Beilstein 04, 405
Merck Index 10, 2948
Solubility Information Solubility in water: freely soluble in water
Formula Weight 140.57
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-2, 3-Diaminopropionic acid monohydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • DL-2,3-Diaminopropionic acid monohydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.