missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DKC1 Recombinant Rabbit Monoclonal Antibody (23GB1810), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553109
This item is not returnable.
View return policy
Description
This gene is a member of the H/ACA snoRNPs gene family. snoRNPs are involved in various aspects of rRNA processing and modification and have been classified into two families: C/D and H/ACA. The H/ACA snoRNPs also include the NOLA1, 2 and 3 proteins. The protein encoded by this gene and the three NOLA proteins localize to the dense fibrillar components of nucleoli and to coiled bodies in the nucleus. Both 18S rRNA production and rRNA pseudouridylation are impaired if any one of the four proteins is depleted. These four H/ACA snoRNP proteins are also components of the telomerase complex. The protein encoded by this gene is related to the Saccharomyces cerevisiae Cbf5p and Drosophila melanogaster Nop60B proteins. The gene lies in a tail-to-tail orientation with the palmitoylated erythrocyte membrane protein gene and is transcribed in a telomere to centromere direction. Both nucleotide substitutions and single trinucleotide repeat polymorphisms have been found in this gene. Mutations in this gene cause X-linked dyskeratosis congenita, a disease resulting in reticulate skin pigmentation, mucosal leukoplakia, nail dystrophy, and progressive bone marrow failure in most cases. Mutations in this gene also cause Hoyeraal-Hreidarsson syndrome, which is a more severe form of dyskeratosis congenita. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| DKC1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DKC1 | |
| BC068171; CBF5; CBF5 homolog; DKC; DKC1; DKCX; dyskeratosis congenita 1, dyskerin; dyskeratosis congenita 1, dyskerin homolog; dyskerin; dyskerin pseudouridine synthase 1; H/ACA ribonucleoprotein complex subunit 4; H/ACA ribonucleoprotein complex subunit DKC1; Nap57; NOLA4; Nopp140-associated protein of 57 kDa; nucleolar protein family A member 4; Nucleolar protein NAP57; snoRNP protein DKC1; XAP101 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170944, 1736, 245474 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1810 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60832, P40615, Q9ESX5 | |
| DKC1 | |
| A synthesized peptide derived from human DKC1 (350-400AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction