missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DKC1 Recombinant Rabbit Monoclonal Antibody (23GB1810), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | DKC1 |
|---|---|
| Clone | 23GB1810 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087694
![]() |
Invitrogen™
MA553108 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088038
|
Invitrogen™
MA553109 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DKC1 Recombinant Monoclonal antibody specifically detects DKC1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| DKC1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O60832, P40615, Q9ESX5 | |
| 170944, 1736, 245474 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1810 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BC068171; CBF5; CBF5 homolog; DKC; DKC1; DKCX; dyskeratosis congenita 1, dyskerin; dyskeratosis congenita 1, dyskerin homolog; dyskerin; dyskerin pseudouridine synthase 1; H/ACA ribonucleoprotein complex subunit 4; H/ACA ribonucleoprotein complex subunit DKC1; Nap57; NOLA4; Nopp140-associated protein of 57 kDa; nucleolar protein family A member 4; Nucleolar protein NAP57; snoRNP protein DKC1; XAP101 | |
| A synthesized peptide derived from human DKC1 (350-400AA). | |
| Primary | |
| -20°C | |
| DKC1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title