Learn More
New Source™ Disposable Chemical Resistant SMS Coveralls Type 5/6
Product Code. 17004287
Change view
Click to view available options
Size:
2X-Large
3X-Large
Large
Medium
X-Large
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17004287 3X-Large Pack of 50
16691793 Medium Pack of 50
16645042 Large Pack of 50
16655042 X-Large Pack of 50
16665042 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17004287 Supplier New Source™ Supplier No. HY9766XXXL
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Disposable chemical resistant coveralls offering protection against solid particulates and liquid splashes and sprays. CE certified as PPE Cat III under Regulation EU 2016/425
  • Conforms to CE Category III Type 5/6
  • Non-woven SMS disposable fabric
  • Protects against chemical and biological hazards
Specifications
Size 3X-Large
Gender Unisex
Material Non-woven SMS
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Flap
Seam Type Stitched
Resistance Particle
Certifications/Compliance CE Cat III PPE meeting: EN ISO 13982-1:2004, EN ISO 13034: 2005 +A1:2009, EN ISO 14325:2004
Description Disposable coverall with elasticated hood, cuffs, waist and ankles. Without shoe cover.
Weight (Metric) 55 gsm