missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Dispense Bottle with Filter, 10L

Product Code. 13593040 Shop All Agilent BioTek Products
Click to view available options
Capacity (Metric):
10 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13593040

Brand: Agilent BioTek 1173000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with 405 LS and 405 TS Microplate Washers

  • Dispense Bottle - with Filter, 10L
  • For use with 405 LS and 405 TS Microplate Washers
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10 L
Type Dispense Bottle
For Use With (Equipment) 405 LS Microplate Washer, 405 Touch Microplate Washer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.