Learn More
BRAND™ Bottletop Dispenser Thread Adapters
Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.Bottletop Dispenser Thread Adapters are available in a variety of thread sizes. For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
Brand: BRAND™ 704281
Code : 03
Additional Details : Weight : 0.05000kg
Description
- Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.
- For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
- Available in a variety of thread sizes.
Specifications
Dispenser Thread Adapter | |
Dispensette Direct Mounting on Drum | |
1 Ea. |
PTFE | |
Dispensette™III, Dispensette™ Organic Remote Dispensing Unit | |
32 GL (inner thread) |