Learn More
BRAND™ Bottletop Dispenser Thread Adapters
Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.Bottletop Dispenser Thread Adapters are available in a variety of thread sizes. For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
£11.50 - £87.70
Product Code | Brand | Material | Thread Size | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Thread Size | For Use With (Equipment) | Price | Quantity & Availability | |||
10373821
|
BRAND™
704338 |
Polypropylene | GL 32/38 | Titrette™ |
£11.50
Each |
In Stock
Log in to see stock. |
|||
10136720
|
BRAND™
704399 |
ETFE | GL 45/38 | Dispensette™, seripettor™, Titrette™, QuikSip™ |
£24.55
Each |
In Stock
Log in to see stock. |
|||
11613179
|
BRAND™
704431 |
Polypropylene | GL 45/35 | Dispensette™, seripettor™, Titrette™, QuikSip™ |
£12.15
Each |
In Stock
Log in to see stock. |
|||
10518781
|
BRAND™
704270 |
Stainless Steel | 2 in. (outer thread), 3/4 in. (inner thread) | Dispensette™III, Dispensette™ Organic Remote Dispensing Unit |
£87.70
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||
10798764
|
BRAND™
704281 |
PTFE | 32 GL (inner thread) | Dispensette™III, Dispensette™ Organic Remote Dispensing Unit |
£47.20
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
10263581
|
BRAND™
704282 |
PTFE | 32 GL (outer thread) | Dispensette™III, Dispensette™ Organic Remote Dispensing Unit |
£30.65
Each |
In Stock
Log in to see stock. |
|||
Description
- Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.
- For use with Dispensette™, seripettor™, Titrette™, QuikSip™ bottletop dispensers.
- Available in a variety of thread sizes.
Specifications
Dispenser Thread Adapter | |
1 Ea. |