Learn More
BRAND™ Bottletop Dispenser Thread Adapters
Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.Bottletop Dispenser Thread Adapters are available in a variety of thread sizes. For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
Brand: BRAND™ 704270
Code : 03
Additional Details : Weight : 0.36000kg
Description
- Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.
- For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
- Available in a variety of thread sizes.
Specifications
Dispenser Thread Adapter | |
Dispensette™III, Dispensette™ Organic Remote Dispensing Unit | |
2 in. (outer thread), 3/4 in. (inner thread) |
Stainless Steel | |
1 Ea. |