missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disc-Shaped PTFE Stir Bars

Product Code. 11878892 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric):
9 mm
10 mm
20 mm
30 mm
Height (Metric):
6mm
10mm
12mm
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Height (Metric) Unit Size
11878892 10 mm 6mm Pack of 10
11888892 20 mm 10mm Pack of 10
11868892 9 mm 6mm Pack of 10
16205841 30 mm 12mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11878892
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Provides stable stirring action for tubes. The disc-shaped stir bars are excellent for applications involving tubes.

  • Stable, magnetic stir bar
  • For use in tubes

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Tubes
Height (Metric) 6mm
Diameter (Metric) 10 mm
Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.