missing translation for 'onlineSavingsMsg'
Learn More

Dirhodium Tetracaprolactamate >90%, TRC

Product Code. 30386548
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386548 50 mg
30375576 5 mg
30380452 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386548 Supplier Toronto Research Chemicals Supplier No. TRCD49495850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138984-26-6
Molecular Formula C6 H10 N O Rh2
Molecular Weight (g/mol) 317.961
Synonym tetrakis[μ-(hexahydro-2H-azepin-2-onato-kN1:kO2)]di-Rhodium (Rh-Rh), tetrakis[μ-(hexahydro-2H-azepin-2-onato-N1:O2)]di-Rhodium (Rh-Rh), Dirhodium(II) caprolactamate
IUPAC Name 8λ3-oxa-1-aza-9λ3-rhodabicyclo[5.2.0]non-7-en-9-ylrhodium
SMILES [Rh][Rh]1N2CCCCCC2=[O]1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H11NO.2Rh/c8-6-4-2-1-3-5-7-6;;/h1-5H2,(H,7,8);;/q;;+1/p-1
Formula Weight 317.887
Chemical Name or Material Dirhodium Tetracaprolactamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.