accessibility menu, dialog, popup

UserName

ONHONHONHONHRhRh

CAS RN 138984-26-6

IUPAC Name: tetrakis(azepan-2-one) dirhodium
Synonyms: tetrakis(caprolactam) dirhodium
Molecular Weight (g/mol): 658.45
Molecular Formula: C24H44N4O4Rh2
InChi Key: SSCZKSBMOZFVCQ-UHFFFAOYSA-N
SMILES: [Rh].[Rh].O=C1CCCCCN1.O=C1CCCCCN1.O=C1CCCCCN1.O=C1CCCCCN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results