missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene glycol monobutyl ether

Product Code. 17241302 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.50 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17241302 1 L 1 L
17251302 2.5 L 2.50 L
17261302 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17241302 Supplier Thermo Scientific Acros Supplier No. 467260010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29911-28-2 | C10H22O3 | 190.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29911-28-2
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
InChI Key CUVLMZNMSPJDON-UHFFFAOYNA-N
IUPAC Name 1-[(1-butoxypropan-2-yl)oxy]propan-2-ol
SMILES CCCCOCC(C)OCC(C)O

Specifications

Melting Point -75.0°C
Density 0.9100 g/mL
Boiling Point 230.0°C
Flash Point 100°C
Quantity 1 L
Specific Gravity 0.91
Solubility Information Solubility aq. soln.: 40-45 g/L (25°C).
Viscosity 4.35 mPa.s (25°C)
Formula Weight 190.28
Chemical Name or Material Dipropylene glycol monobutyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.