accessibility menu, dialog, popup

UserName

H3COCH3OCH3OH

CAS RN 29911-28-2

IUPAC Name: 1-[(1-butoxypropan-2-yl)oxy]propan-2-ol
Synonyms: ppg-2 butyl ether ppg-4 butyl ether ppg-5 butyl ether ppg-9 butyl ether ppg-12 butyl ether ppg-14 butyl ether ppg-15 butyl ether ppg-16 butyl ether ppg-17 butyl ether ppg-18 butyl ether
Molecular Weight (g/mol): 190.28
Molecular Formula: C10H22O3
InChi Key: CUVLMZNMSPJDON-UHFFFAOYNA-N
SMILES: CCCCOCC(C)OCC(C)O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results