missing translation for 'onlineSavingsMsg'
Learn More

Dipropyl Sodium Phosphate, TRC

Product Code. 30388277
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388277 50 mg
30383968 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388277 Supplier Toronto Research Chemicals Supplier No. TRCD49239050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4545-54-4
Molecular Formula C6 H14 Na O4 P
Molecular Weight (g/mol) 204.137
Synonym Dipropyl Ester Phosphoric Acid Sodium Salt
IUPAC Name dipropoxyphosphoryloxysodium
SMILES CCCOP(=O)(O[Na])OCCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H15O4P.Na/c1-3-5-9-11(7,8)10-6-4-2;/h3-6H2,1-2H3,(H,7,8);/q;+1/p-1
Formula Weight 204.053
Chemical Name or Material Dipropyl Sodium Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.