CAS RN 4545-54-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dipropyl Sodium Phosphate, TRC
CAS: 4545-54-4 Molecular Formula: C6 H14 Na O4 P Molecular Weight (g/mol): 204.137 Synonym: Dipropyl Ester Phosphoric Acid Sodium Salt IUPAC Name: dipropoxyphosphoryloxysodium SMILES: CCCOP(=O)(O[Na])OCCC