Learn More
Diphenylphosphonic azide, 97%, Thermo Scientific Chemicals
Store cold
£22.70 - £135.00
Chemical Identifiers
CAS | 26386-88-9 |
---|---|
Molecular Formula | C12H10N3O3P |
Molecular Weight (g/mol) | 275.20 |
MDL Number | MFCD00001987 |
InChI Key | SORGEQQSQGNZFI-UHFFFAOYSA-N |
Synonym | diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av |
PubChem CID | 123414 |
IUPAC Name | [azido(phenoxy)phosphoryl]oxybenzene |
SMILES | [N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410073
|
Thermo Scientific Alfa Aesar
A12124.06 |
5 g |
£22.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420073
|
Thermo Scientific Alfa Aesar
A12124.14 |
25 g |
£45.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430073
|
Thermo Scientific Alfa Aesar
A12124.22 |
100 g |
£135.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Diphenylphosphonic azide acts as a reagent for the synthesis of peptides and phosphoramidates by reacting with amines. It is also used in the preparation of oligosaccharides linked with carbamate and urea bonds utilizing modified Curtis rearrangement. It is involved in pseudohalogen replacement of the azido group by treatment with nucleophilic reagents, such as water, butanol, ammonia, and various amines. Further, it is used as a hydroazidation catalyst for preparation of organoazides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenylphosphonic azide acts as a reagent for the synthesis of peptides and phosphoramidates by reacting with amines. It is also used in the preparation of oligosaccharides linked with carbamate and urea bonds utilizing modified Curtis rearrangement. It is involved in pseudohalogen replacement of the azido group by treatment with nucleophilic reagents, such as water, butanol, ammonia, and various amines. Further, it is used as a hydroazidation catalyst for preparation of organoazides.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with acids and oxidizing agents.
Chemical Identifiers
26386-88-9 | |
275.20 | |
SORGEQQSQGNZFI-UHFFFAOYSA-N | |
123414 | |
[N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1 |
C12H10N3O3P | |
MFCD00001987 | |
diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av | |
[azido(phenoxy)phosphoryl]oxybenzene |
Specifications
26386-88-9 | |
161°C to 162°C (0.5 mmHg) | |
97% | |
1.552 | |
UN3278 | |
diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av | |
SORGEQQSQGNZFI-UHFFFAOYSA-N | |
[azido(phenoxy)phosphoryl]oxybenzene | |
123414 | |
97% | |
Diphenylphosphonic azide |
1.275 | |
>110°C (230°F) | |
C12H10N3O3P | |
MFCD00001987 | |
2058967 | |
Immiscible with water. | |
[N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1 | |
275.20 | |
275.2 | |
Liquid |
Safety and Handling
GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
H301-H310+H330-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 247-644-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only