missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphonic azide, 97%

Product Code. 11410073
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410073 5 g 5g
11420073 25 g 25g
11430073 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410073 Supplier Thermo Scientific Alfa Aesar Supplier No. A12124.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Store cold | CAS: 26386-88-9 | C12H10N3O3P | 275.20 g/mol

Diphenylphosphonic azide acts as a reagent for the synthesis of peptides and phosphoramidates by reacting with amines. It is also used in the preparation of oligosaccharides linked with carbamate and urea bonds utilizing modified Curtis rearrangement. It is involved in pseudohalogen replacement of the azido group by treatment with nucleophilic reagents, such as water, butanol, ammonia, and various amines. Further, it is used as a hydroazidation catalyst for preparation of organoazides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphonic azide acts as a reagent for the synthesis of peptides and phosphoramidates by reacting with amines. It is also used in the preparation of oligosaccharides linked with carbamate and urea bonds utilizing modified Curtis rearrangement. It is involved in pseudohalogen replacement of the azido group by treatment with nucleophilic reagents, such as water, butanol, ammonia, and various amines. Further, it is used as a hydroazidation catalyst for preparation of organoazides.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26386-88-9
Molecular Formula C12H10N3O3P
Molecular Weight (g/mol) 275.20
MDL Number MFCD00001987
InChI Key SORGEQQSQGNZFI-UHFFFAOYSA-N
Synonym diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av
PubChem CID 123414
IUPAC Name [azido(phenoxy)phosphoryl]oxybenzene
SMILES [N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Density 1.275
Boiling Point 161°C to 162°C (0.5 mmHg)
Flash Point >110°C (230°F)
Assay Percent Range 97%
Refractive Index 1.552
Quantity 5 g
UN Number UN3278
Beilstein 2058967
Solubility Information Immiscible with water.
Formula Weight 275.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diphenylphosphonic azide
Show More Show Less
compliance-icons
Product Identifier
  • Diphenylphosphonic azide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Phosphorazidic acid,diphenyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.