missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Visocolor™ ECO Chloride Colorimetric Test Kit
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
1 Each
Unit Size:
65g
Specifications
Specifications
| For Use With (Application) | Chloride ions react with mercuric thiocyanate to produce undissociated mercuric chloride and to liberate thiocyanate ions |
| Quantity | 1 Each |
| Range | 1 to 60mg/L C |
| Sufficient For | 90 Determinations |
| Type | ECO Chloride Colorimetric Test Kit |
Safety and Handling
Product Identifier
- VISOCOLOUR ECO Chloride
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 4
- Substances and mixtures corrosive to metals Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 3
- Acute toxicity Category 4
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H290-May be corrosive to metals.
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H312-Harmful in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- H413-May cause long lasting harmful effects to aquatic life.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- MIXTURE LIST-Contains : Nitric acid, Iron(III) nitrate, Mercury(II) thiocyanate, Methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction