missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Diphenyl Blue

Product Code. 16471198
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471198 Supplier MedChemExpress Supplier No. HYD0970
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Diphenyl Blue (Direct Blue 14) is used as reference dye for group of azo dye.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 5 g
Shipping Condition Room temperature
Solubility H2O : 10 mg/mL (10.41 mM; Need ultrasonic)
Target HBV
Molecular Weight (g/mol) 960.81 g/mol
CAS 72-57-1
Synonym Direct Blue 14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.