missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIO2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26513-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the iodothyronine deiodinase family. It activates thyroid hormone by converting the prohormone thyroxine by outer ring deiodination to bioactive 3,3′,5-triiodothyronine residues encoded by the UGA codon, which normally signals translation termination. The 3′ UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence , which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| DIO2 | |
| Polyclonal | |
| Unconjugated | |
| DIO2 | |
| 5DII, D2, DIO2, DIOII, ITDI2, SelY, TXDI2, Type 2 DI, Type II 5 deiodinase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13371, 1734 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92813, Q9Z1Y9 | |
| DIO2 | |
| DIO2 Fusion Protein Ag24119 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction