missing translation for 'onlineSavingsMsg'
Learn More

Dimethylarsinothioic Acid, TRC

Product Code. 30180044
Change view
Click to view available options
Quantity:
10 mg
250 mg
1 g
Unit Size:
10mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180044 250 mg 250mg
30180087 1 g 1g
30180045 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180044 Supplier Toronto Research Chemicals Supplier No. TRCD471360250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754217-65-7
Molecular Formula C2H7AsOS
Molecular Weight (g/mol) 154.06
Synonym Dimethylarsinothioic O-Acid;
IUPAC Name dimethylarsinothioic S-acid
SMILES O=[As](S)(C)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C2H7AsOS/c1-3(2,4)5/h1-2H3,(H,4,5)
Formula Weight 153.94
Chemical Name or Material Dimethylarsinothioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.